Anti-S6K1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX14708
89360-052EA 626.11 USD
89360-052
Anti-S6K1 Goat Polyclonal Antibody
Antibodies
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized.

Approx 65-70kDa band observed in Human Skeletal Muscle and Placenta lysates (predicted MW of 60kDa according to NP_003152.1). Recommended for use at 0.2-1µg/ml.

Type: Primary
Antigen: S6K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR