Anti-ALB Mouse Monoclonal Antibody [clone: 1A9]

Supplier: GeneTex
GTX10242
89350-734EA 717.07 USD
89350-734
Anti-ALB Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Mouse monoclonal antibody [1A9] to Albumin

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 65 kDa. Can be paired for ELISA with GTX10241 Mouse monoclonal [15C7] to Albumin . Produces sensitive assay system with rapid kinetics (10-15 mins in plate format) for the detection of albumin in human urine. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR