Anti-CYP4A11 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103518
89350-024EA 600.25 USD
89350-024
Anti-CYP4A11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CYP4A11 (cytochrome P450, family 4, subfamily A, polypeptide 11)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYP4A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR