Anti-STIM1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-60547SS NB110-60547
102124-224EA 208 USD
102124-224 102124-210
Anti-STIM1 Rabbit Polyclonal Antibody
Antibodies
The STIM1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STIM1. This antibody reacts with human, mouse, rat, bovine, canine. The STIM1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: STIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR