Anti-RCAN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46852SS NBP1-46852
102176-604EA 208 USD
102176-604 102176-590
Anti-RCAN1 Rabbit Polyclonal Antibody
Antibodies
The RCAN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RCAN1. This antibody reacts with human, mouse. The RCAN1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000

Type: Primary
Antigen: RCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR