Anti-AG-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-05936 NBP1-05936SS
102156-926EA 503.38 USD
102156-926 102156-940
Anti-AG-2 Rabbit Polyclonal Antibody
Antibodies
The AG-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AG-2. This antibody reacts with human, mouse, bovine. The AG-2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: AG-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR