Anti-SIAH2 Mouse Monoclonal Antibody [clone: 24E6H3]

Supplier: Novus Biologicals
NB110-88113SS NB110-88113
102126-086EA 208 USD
102126-086 102126-084
Anti-SIAH2 Mouse Monoclonal Antibody [clone: 24E6H3]
Antibodies
The SIAH2 Antibody (24E6H3) from Novus Biologicals is a mouse monoclonal antibody to SIAH2. This antibody reacts with human, porcine, drosophila, mouse (negative). The SIAH2 Antibody (24E6H3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Type: Primary
Antigen: SIAH2
Clonality: Monoclonal
Clone: 24E6H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Drosophila, Mouse (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR