Anti-GFAP Mouse Monoclonal Antibody [clone: 5C10]

Supplier: Novus Biologicals
NBP1-05197 NBP1-05197SS
102156-688EA 601.85 USD
102156-688 102156-690
Anti-GFAP Mouse Monoclonal Antibody [clone: 5C10]
Antibodies
The GFAP Antibody (5C10) from Novus Biologicals is a mouse monoclonal antibody to GFAP. This antibody reacts with human, mouse, rat, porcine, avian, bovine, chicken, equine, mammal. The GFAP Antibody (5C10) has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:500 - 1:1000

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 5C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Chicken, Equine, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR