Anti-SR-BI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB400-113 NB400-113SS
102131-382EA 503.38 USD
102131-382 102131-384
Anti-SR-BI Rabbit Polyclonal Antibody
Antibodies
The SR-BI Antibody from Novus Biologicals is a rabbit polyclonal antibody to SR-BI. This antibody reacts with human, mouse, rat. The SR-BI Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Blocking / Neutralizing.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: SR-BI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR