Anti-EN-RAGE/S100A12 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-90136 NB110-90136SS
102126-164EA 503.38 USD
102126-164 102126-166
Anti-EN-RAGE/S100A12 Rabbit Polyclonal Antibody
Antibodies
The EN-RAGE / S100A12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EN-RAGE / S100A12. This antibody reacts with human, mouse (negative), rat (negative). The EN-RAGE / S100A12 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: EN-RAGE/S100A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse (Negative), Rat (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR