Anti-KI67 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-90592SS NB110-90592
102126-170EA 208 USD
102126-170 102126-168
Anti-KI67 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Ki67 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:3200

Type: Primary
Antigen: KI67
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR