3-Aminobenzylamine
Supplier: Matrix Scientific
Synonyms:
alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline
056887-5G
056887-25G
101780-678EA
73.85
USD
101780-678
101780-680
3-Aminobenzylamine
3-Aminobenzylamine
Hazard: Corrosive
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Melting Pt: 41…45 °C |
MDL Number:
MFCD00078355 CAS Number: 4403-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...