6-Amino-3-cyano-2-methylpyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile
020969-1G
020969-5G
101780-264EA
169.23
USD
101780-264
101780-266
6-Amino-3-cyano-2-methylpyridine ≥98%
6-Amino-3-cyano-2-methylpyridine
Hazard: Toxic by inhalation/Irritant
Formula:
C₇H₇N₃ Melting Pt: 168…170 °C |
MDL Number:
MFCD05865116 CAS Number: 183428-90-2 |
Specification Test Results
Melting point | 168-170° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...