4-Iodophenylhydrazine ≥96%

Supplier: Matrix Scientific

007214-1G 007214-5G
101779-492EA 115.38 USD
101779-492 101779-494
4-Iodophenylhydrazine ≥96%
4-Iodophenylhydrazine

Hazard: Toxic/Store in Freezer/Light Sensitive

Formula: C₆H₇IN₂
Melting Pt: 102…105 °C
MDL Number: MFCD00052688
CAS Number: 13116-27-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 102-105°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR