2-Amino-6-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
007185-1G
007185-5G
101779-462EA
26.15
USD
101779-462
101779-464
2-Amino-6-(trifluoromethyl)pyridine ≥97%
2-Amino-6-(trifluoromethyl)pyridine
Hazard: Toxic/Irritant
Formula:
C₆H₅F₃N₂ Melting Pt: 88…89 °C |
MDL Number:
MFCD03093868 CAS Number: 34486-24-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 88-89° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...