Silver tetrafluoroborate ≥99%
Supplier: Matrix Scientific
Synonyms:
Argentous tetrafluoroborate
006354-5G
006354-25G
101779-030EA
64.62
USD
101779-030
101779-032
Silver tetrafluoroborate ≥99%
Silver tetrafluoroborate
TCA listed: T
Hazard: Corrosive/Moisture Sensitive
Formula:
AgBF₄ Melting Pt: 200 °C |
MDL Number:
MFCD00003408 CAS Number: 14104-20-2 UN: 3260 ADR: 8,II |
Specification Test Results
Melting point | 200° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...