2,4,6-Tris(trifluoromethyl)-1,3,5-triazine ≥99%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Tris(trifluoromethyl)-s-triazine

006183-1G 006183-5G
101778-926EA 83.08 USD
101778-926 101778-928
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine ≥99%
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine

Hazard: Irritant

Formula: C₆F₉N₃
MW: 285.07
Boiling Pt: 95 °C
MDL Number: MFCD00042436
CAS Number: 368-66-1

Order Now

Specification Test Results

Boiling point 95°
ND20 1.3231


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR