α'-Bromo-α,α,α-trifluoro-p-xylene ≥98%

Supplier: Matrix Scientific

005486-100G 005486-25G
101778-394EA 144.62 USD
101778-394 101778-392
α'-Bromo-α,α,α-trifluoro-p-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-p-xylene

Hazard: Corrosive/Irritant/Lachrymator/Store in Freezer

Formula: C₈H₆BrF₃
Boiling Pt: 65…66 °C (5 mmHg)
Melting Pt: 30…31 °C
Density: 1.546
MDL Number: MFCD00000403
CAS Number: 402-49-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 65-66°/5mm
Density 1.546
ND20 1.484

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR