3,5-Dichlorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

005175-25G 005175-5G
101778-048EA 107.69 USD
101778-048 101778-046
3,5-Dichlorobenzenesulfonyl chloride ≥98%
3,5-Dichlorobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₃Cl₃O₂S
Melting Pt: 30…32 °C
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 30-32°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR