ω,4-Dibromoacetophenone ≥98%

Supplier: Matrix Scientific

004903-100G 004903-25G
101777-840EA 55.38 USD
101777-840 101777-838
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone

TCA listed: T
Hazard: Corrosive/Lachrymator

Formula: C₈H₆Br₂O
Melting Pt: 108…110 °C
MDL Number: MFCD00000200
CAS Number: 99-73-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 108-110°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR