2,2,2-Trifluoroethyl acrylate ≥99%

Supplier: Matrix Scientific

Synonyms: 2,2,2-TrifluoroethylacrylateAcrylic acid 2,2,2-trifluoroethyl ester

003713-100G 003713-25G
101777-058EA 196.92 USD
101777-058 101777-056
2,2,2-Trifluoroethyl acrylate ≥99%
2,2,2-Trifluoroethyl acrylate

TCA listed: T
Hazard: Flammable/Irritant/Refrigerate

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 46 °C (125 mmHg)
Density: 1.216
Flash Pt: 12 °C
MDL Number: MFCD00000444
CAS Number: 407-47-6
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Boiling point 46°/125mm
Density 1.216
ND20 1.35

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR