2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%

Supplier: Matrix Scientific

Synonyms: 1H,1H-Perfluoro-1-butanol2,2,3,3,4,4,4-Heptafluorobutan-1-olHeptafluoro-1-butanol

001211-100G 001211-25G
101775-248EA 146.15 USD
101775-248 101775-250
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol

TCA listed: T
Hazard: Flammable

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 96…97 °C
Density: 1.6
MDL Number: MFCD00004674
CAS Number: 375-01-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Boiling point 96-97°
Density 1.6
ND20 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR