2,4,5-Trifluorobenzonitrile ≥98%

Supplier: Matrix Scientific

003628-5G 003628-25G
101776-908EA 49.23 USD
101776-908 101776-910
2,4,5-Trifluorobenzonitrile ≥98%
2,4,5-Trifluorobenzonitrile

Hazard: Toxic

Formula: C₇H₂F₃N
Density: 1.373
MDL Number: MFCD00013289
CAS Number: 98349-22-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.373
ND20 1.4736


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR