Silver trifluoroacetate ≥98%

Supplier: Matrix Scientific

Synonyms: Silver(I) trifluoroacetate

002912-5G 002912-25G
101776-426EA 84.62 USD
101776-426 101776-428
Silver trifluoroacetate ≥98%
Silver trifluoroacetate

TCA listed: T
Hazard: Toxic/Light Sensitive/Hygroscopic

Formula: C₂AgF₃O₂
MW: 220.88
Melting Pt: 257…260 °C
MDL Number: MFCD00013199
CAS Number: 2966-50-9

Order Now

Specification Test Results

Melting point 257-260°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR