2-Amino-6-fluorobenzonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-3-fluoroaniline

001697-25G 001697-100G
101775-510EA 18.46 USD
101775-510 101775-512
2-Amino-6-fluorobenzonitrile ≥99%
2-Amino-6-fluorobenzonitrile

Hazard: Toxic

Formula: C₇H₅FN₂
MW: 136.13
Boiling Pt: 272
Melting Pt: 127
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Melting point 125-127°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR