Deuterium oxide ≥99.95% (Isotopic), MagniSolv™ for NMR spectroscopy pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Heavy water
, Water-D2
M-Clarity™ quality level = MQ200
1.03428.0100
1.03428.0005
1.03428.0010
1.03428.0005
1.03428.0009
1.03428.0010
1.03428.0009
1.03428.0100
EM1.03428.0005EA
100
USD
CA1.03428.0100
EM1.03428.0005
CA1.03428.0010
CA1.03428.0005
EM1.03428.0009
CA1.03428.0009
EM1.03428.0010
EM1.03428.0100
Deuterium oxide ≥99.95% (Isotopic), MagniSolv™ for NMR spectroscopy pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Deuterium oxide
Formula:
D₂O MW: 20 g/mol Boiling Pt: 101.3 °C (1013 hPa) Melting Pt: 3.8 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044636 CAS Number: 7789-20-0 EINECS: 232-148-9 Merck Index: 12,02984 |
Specification Test Results
Deuteration degree | ≥ 99.95 % |
Performance test (NMR-spectrum) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...