Nitric acid 69.5%, Cleanroom® LP for the electronics industry
Supplier: KMG
|
Danger
|
200287
71009-340CS
954.61
USD
71009-340
Nitric acid 69.5%, Cleanroom® LP for the electronics industry
Nitric acid
Nitric acid is a strong oxidizing agent. Nitric acid is used to oxidize silicon to silicon dioxide and to dissolve metals to metal nitrates. Nitric acid is used in combination with HF in mixed acid etchants to etch silicon.
Storage: <75⁰ F
Formula:
HNO₃ MW: 63.01 g/mol Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Specification Test Results
Assay (HNO3) | 69.00 to 70.00 % |
Color | 10 APHA |
Appearance | Conforms |
Chloride (Cl) | 80 ppb |
Phosphate (PO4) | 200 ppb |
Sulfate (SO4) | 200 ppb |
Aluminum (Al) | 20.00 ppb |
Antimony (Sb) | 2.00 ppb |
Arsenic (As) | 2.00 ppb |
Barium (Ba) | 10.00 ppb |
Beryllium (Be) | 10.00 ppb |
Bismuth (Bi) | 20.00 ppb |
Boron (B) | 10.00 ppb |
Cadmium (Cd) | 20.00 ppb |
Calcium (Ca) | 20.00 ppb |
Chromium (Cr) | 20.00 ppb |
Cobalt (Co) | 10.00 ppb |
Copper (Cu) | 20.00 ppb |
Gallium (Ga) | 10.00 ppb |
Germanium (Ge) | 10.00 ppb |
Gold (Au) | 20.00 ppb |
Iron (Fe) | 20.00 ppb |
Lead (Pb) | 20.00 ppb |
Lithium (Li) | 10.00 ppb |
Magnesium (Mg) | 20.00 ppb |
Manganese (Mn) | 10.00 ppb |
Nickel (Ni) | 20.00 ppb |
Potassium (K) | 20.00 ppb |
Silicon (Si) | 30.00 ppb |
Silver (Ag) | 20.00 ppb |
Sodium (Na) | 20.00 ppb |
Strontium (Sr) | 20.00 ppb |
Tantalum (Ta) | 10.00 ppb |
Thallium (Tl) | 10.00 ppb |
Tin (Sn) | 20.00 ppb |
Titanium (Ti) | 10.00 ppb |
Tungsten (W) | 500.00 ppb |
Vanadium (V) | 10.00 ppb |
Zinc (Zn) | 20.00 ppb |
Zirconium (Zr) | 10.00 ppb |
0.5μ Particle Count | 60 par/ml |
1.0μ Particle Count | 10 par/ml |
10 par/ml |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...