Hydrogen peroxide 30%, Cleanroom® MB for the electronics industry
Supplier: KMG
|
Danger
|
64025
71009-334CS
657.83
USD
71009-334
Hydrogen peroxide 30%, Cleanroom® MB for the electronics industry
Hydrogen peroxide
Hydrogen peroxide is used in SC-1 and SC-2 solutions to remove particles from silicon wafers and to remove surface metallic contamination. The purity of the hydrogen peroxide is critical to surface roughness and metallic residual levels on the wafer.
Formula:
H₂O₂ MW: 34.01 g/mol Boiling Pt: ∼107 °C (1013 hPa) Melting Pt: < 0 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1,II |
Specification Test Results
Assay (H2O2) | 30.00 to 32.00 % |
Assay (H2O) | 68.00 to 70.00 % |
Color | 6 APHA |
Appearance | Conforms |
Free Acid | 0.60 μeq/g |
Total Organic Carbon | 20,000 ppb |
Chloride (Cl) | 60 ppb |
Nitrate (NO3) | 200 ppb |
Phosphate (PO4) | 200 ppb |
Sulfate (SO4) | 200 ppb |
Aluminum (Al) | 10.0 ppb |
Antimony (Sb) | 5.0 ppb |
Arsenic (As) | 5.0 ppb |
Barium (Ba) | 10.0 ppb |
Beryllium (Be) | 10.0 ppb |
Bismuth (Bi) | 10.0 ppb |
Boron (B) | 10.0 ppb |
Cadmium (Cd) | 10.0 ppb |
Calcium (Ca) | 10.0 ppb |
Chromium (Cr) | 10.0 ppb |
Cobalt (Co) | 10.0 ppb |
Copper (Cu) | 10.0 ppb |
Gallium (Ga) | 10.0 ppb |
Germanium (Ge) | 10.0 ppb |
Gold (Au) | 10.0 ppb |
Iron (Fe) | 10.0 ppb |
Lead (Pb) | 10.0 ppb |
Lithium (Li) | 10.0 ppb |
Magnesium (Mg) | 10.0 ppb |
Manganese (Mn) | 10.0 ppb |
Molybdenum (Mo) | 10.0 ppb |
Nickel (Ni) | 10.0 ppb |
Niobium (Nb) | 10.0 ppb |
Potassium (K) | 10.0 ppb |
Silicon (Si) | 10.0 ppb |
Silver (Ag) | 10.0 ppb |
Sodium (Na) | 10.0 ppb |
Strontium (Sr) | 10.0 ppb |
Tantalum (Ta) | 10.0 ppb |
Thallium (Tl) | 10.0 ppb |
Tin (Sn) | 10.0 ppb |
Titanium (Ti) | 10.0 ppb |
Vanadium (V) | 10.0 ppb |
Zinc (Zn) | 10.0 ppb |
Zirconium (Zr) | 10.0 ppb |
0.2μ Particle Count | 1,000 par/ml |
0.3μ Particle Count | 1,000 par/ml |
0.5μ Particle Count | 25 par/ml |
1.0μ Particle Count | 10 par/ml |
10 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...