Hydrofluoric acid 49%, Cleanroom® MB MB for the electronics industry

Supplier: KMG

64003 64004
71009-330CS 831.53 USD
71009-330 71009-362
Hydrofluoric acid 49%, Cleanroom® MB MB for the electronics industry
Hydrofluoric acid

Hydrofluoric Acid 49% is used to soluabilize many oxides, particularly silicon dioxide, the most widely used insulator on semiconductor devices.

Formula: HF
MDL Number: MFCD00011346
CAS Number: 7664-39-3
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Assay (HF) 48.80 to 49.20 %
Color 10 APHA
Appearance Conforms
Fluosilicic Acid 50 ppm
Chloride (Cl) 2,000 ppb
Nitrate (NO3) 1,000 ppb
Phosphate (PO4) 400 ppb
Sulfate (SO4) 500 ppb
Aluminum (Al) 10.0 ppb
Antimony (Sb) 5.0 ppb
Arsenic (As) 5.0 ppb
Barium (Ba) 10.0 ppb
Beryllium (Be) 5.0 ppb
Bismuth (Bi) 5.0 ppb
Boron (B) 10.0 ppb
Cadmium (Cd) 10.0 ppb
Calcium (Ca) 10.0 ppb
Chromium (Cr) 10.0 ppb
Cobalt (Co) 10.0 ppb
Copper (Cu) 10.0 ppb
Gallium (Ga) 10.0 ppb
Germanium (Ge) 10.0 ppb
Gold (Au) 5.0 ppb
Iron (Fe) 10.0 ppb
Lead (Pb) 10.0 ppb
Lithium (Li) 5.0 ppb
Magnesium (Mg) 10.0 ppb
Manganese (Mn) 10.0 ppb
Molybdenum (Mo) 10.0 ppb
Nickel (Ni) 10.0 ppb
Niobium (Nb) 10.0 ppb
Potassium (K) 10.0 ppb
Silver (Ag) 5.0 ppb
Sodium (Na) 10.0 ppb
Strontium (Sr) 10.0 ppb
Tantalum (Ta) 10.0 ppb
Thallium (Tl) 10.0 ppb
Tin (Sn) 10.0 ppb
Titanium (Ti) 10.0 ppb
Vanadium (V) 10.0 ppb
Zinc (Zn) 10.0 ppb
Zirconium (Zr) 10.0 ppb
0.5μ Particle Count 10 par/ml
1.0μ Particle Count 2 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR