Ethanol, OmniPur®, pure (200 Proof), Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl alcohol

4450-500ML 4455
EM-4450SEA 98.67 USD
EM-4450S EM-4455S
Ethanol, OmniPur®, pure (200 Proof), Sigma-Aldrich®
Ethanol
Formula: C₂H₅OH
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114.5 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR