Choline hydrogen tartrate 98 FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

C1228-125GM C1228-500GM C1228-12KG C1228-50KGBL C1228-2.5KG
95032-556EA 102.94 USD
95032-556 95032-560 700007-514 700007-516 95032-558
Choline hydrogen tartrate 98 FCC suitable for use as excipient
Choline hydrogen tartrate

Choline Bitartrate, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₉H₁₉NO₇
MW: 253.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036332
CAS Number: 87-67-2
EINECS: 201-763-4

Order Now

Specification Test Results

Assay (%) 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR