N-Acetyl-β-D-glucosamine ≥99%, Reagent Grade for biochemistry
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
A1269-25KGBL
A1269-500GM
A1269-25GM
700008-458EA
5052.05
USD
700008-458
700008-460
700008-456
N-Acetyl-β-D-glucosamine ≥99%, Reagent Grade for biochemistry
N-Acetyl-β-D-glucosamine
N-Acetyl-D-glucosamine, Reagent makes up the outer layers of crustaceans and insects and is the main component of the cell walls of most fungi. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₈H₁₅NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00136044 CAS Number: 7512-17-6 EINECS: 231-368-2 |
Specification Test Results
Assay (%) | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...