1-Vinyl-2-pyrrolidone ≥99.0% stabilized
Supplier: TCI America
Synonyms:
N-Vinyl-2-pyrrolidinone
, N-Vinyl-2-pyrrolidone
, 1-Vinyl-2-pyrrolidinone
(stabilized with N,N'-Di-sec-butyl-p-phenylenediamine)
CAS Number: 88-12-0
MDL Number: MFCD00003197
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 148
Flash Point (°C): 102
Freezing point (°C): 13
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C
Formula:
C₆H₉NO MW: 111.1418 g/mol Boiling Pt: 148 °C Density: 1.05 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00003197 CAS Number: 88-12-0 EINECS: 201-800-4 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...