Tamoxifen citrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate
CAS Number: 54965-24-1
MDL Number: MFCD00058321
Molecular Formula: C26H29NO
Molecular Weight: 563.65
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 142
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antitumor Ingredients, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate
Formula:
C₂₆H₂₉NO MW: 563.65 g/mol Melting Pt: 142 °C |
MDL Number:
MFCD00058321 CAS Number: 54965-24-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...