Methylene blue ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Methylthioninium chloride 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

A5105-1G
TCA5105-001GEA 41.41 USD
TCA5105-001G
Methylene blue ≥98.0% (by titrimetric analysis)
Methylene blue

[Ion association reagent for spectrophotometric analysis]
CAS Number: 61-73-4
MDL Number: MFCD00012111
Molecular Formula: C16H18ClN3S
Molecular Weight: 319.85
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Lambda max.: 663 nm (EtOH(50vol%))


Product Categories: Bioscience, Stains & Dyes, Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Basic Blue 9

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180 °C
Density: 1.757 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
Merck Index: 12,06137
https://us.vwr.com/stibo/web/std.lang.all/20/78/28932078.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR