Triarachidin ≥98.0%

Supplier: TCI America

Synonyms: Glycerol TrieicosanoateTriarachin1,2,3-Trieicosanoyl Glycerol

T1389-5G T1389-1G
TCT1389-5GEA 290.2 USD
TCT1389-5G TCT1389-001G
Triarachidin ≥98.0%
Triarachidin

CAS Number: 620-64-4
MDL Number: MFCD00042879
Molecular Formula: C63H122O6
Molecular Weight: 975.66
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 78


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Trieicosanoate, Triarachin, 1,2,3-Trieicosanoyl Glycerol

Formula: C₆₃H₁₂₂O₆
MW: 975.66 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00042879
CAS Number: 620-64-4
https://us.vwr.com/stibo/web/std.lang.all/54/12/28995412.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR