Erythrosin B disodium salt ≥95.0% (by UV method)
Supplier: TCI America
|
Warning
|
Synonyms:
Acid Red 51
CAS Number: 16423-68-0
MDL Number: MFCD00144257
Molecular Formula: C20H8I4O5
Molecular Weight: 879.86
Purity/Analysis Method:
Form: Crystal
Color: Yellow Red
Lambda max.: 526 nm (0.02mol/L AcONH4 sol.)
Product Categories: Materials Chemistry, Xanthene Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Iodoeosine Sodium Salt, Tetraiodofluorescein Sodium Salt
Formula:
C₂₀H₆I₄Na₂O₅ MW: 879.86 g/mol |
MDL Number:
MFCD00144257 CAS Number: 16423-68-0 EINECS: 240-474-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...