Toluenediisocyanate mixture of isomers ≥98.0% (2,4-80%, 2,6-20%) for synthesis

Supplier: TCI America
Danger

Synonyms: Toluylene diisocyanate

T0264-500G T0264-25G
TCT0264-500GEA 84.4 USD
TCT0264-500G TCT0264-025G
Toluenediisocyanate mixture of isomers ≥98.0% (2,4-80%, 2,6-20%) for synthesis
Toluenediisocyanate

Form: Clear Liquid
Color: Colorless
Boiling point (°C): 251
Melting point (°C): 14
Flash Point (°C): 137
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bioscience, Reagents for Inflammation & Immunity Disorder Models
Other Product Names / Synonyms: Toluene Diisocyanate

Formula: C₉H₆N₂O₂
Boiling Pt: 251 °C
Melting Pt: 14 °C
Density: 1.22 (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD01778251
CAS Number: 26471-62-5
EINECS: 247-722-4
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR