Sulfaguanidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 57-67-0
MDL Number: MFCD00038136
Molecular Formula: C7H10N4O2S
Molecular Weight: 214.24
Purity/Analysis Method:
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4-Amino-N-amidinobenzenesulfonamide, 4-Aminobenzenesulfonylguanidine, 4-Amino-N-carbamimidoylbenzenesulfonamide
Formula:
C₇H₁₀N₄O₂S Melting Pt: 189 °C |
MDL Number:
MFCD00038136 CAS Number: 57-67-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...