4-Methylumbelliferyl phosphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Methylumbelliferyl phosphoric acid
, Phosphoric Acid 4-Methylumbelliferyl Ester
, 4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphate
CAS Number: 3368-04-5
MDL Number: MFCD00016969
Molecular Formula: C10H9O6P
Molecular Weight: 256.15
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 212
Storage Temperature:
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Phosphoric Acid 4-Methylumbelliferyl Ester
Formula:
C₁₀H₉O₆P MW: 256.15 g/mol Melting Pt: 212 °C |
MDL Number:
MFCD00016969 CAS Number: 3368-04-5 EINECS: 222-137-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...