Pectin (from citrus)
Supplier: TCI America
P0024-25G
P0024-500G
TCP0024-025GEA
39.73
USD
TCP0024-025G
TCP0024-500G
Pectin (from citrus)
Pectin
CAS Number: 9000-69-5
MDL Number: MFCD00081838
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
MDL Number:
MFCD00081838 CAS Number: 9000-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...