4-Nitrophthalhydrazide ≥98.0%

Supplier: TCI America

Synonyms: 4-Nitrophthalhydrazide, 2,3-Dihydro-6-nitro-1,4-phthalazinedione, 4-Nitrophthalic Hydrazide

N0799-5G N0799-25G
TCN0799-5GEA 68 USD
TCN0799-5G TCN0799-25G
4-Nitrophthalhydrazide ≥98.0%
4-Nitrophthalhydrazide

CAS Number: 3682-19-7 MDL Number: MFCD00006891 Molecular Formula: C8H5N3O4 Molecular Weight: 207.15 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 300


Other Product Names / Synonyms: 4-Nitrophthalhydrazide, 2,3-Dihydro-6-nitro-1,4-phthalazinedione

Formula: C₈H₅N₃O₄
Melting Pt: 300 °C
MDL Number: MFCD00006891
CAS Number: 3682-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54f4b3f901941fe948538aa5a1e9d2a2a71e09ef&vid=54f4b3f901941fe948538aa5a1e9d2a2a71e09ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR