4-(4-Aminophenyl)thiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

Synonyms: 4-Thiomorpholinoaniline 1',1'-Dioxide, 4-(4-Aminophenyl)thiomorpholine 1,1-dioxide, 4-(1,1-Dioxothiomorpholino)aniline

A1669-1G A1669-5G
TCA1669-1GEA 58.6 USD
TCA1669-1G TCA1669-5G
4-(4-Aminophenyl)thiomorpholine-1,1-dioxide ≥98.0%
4-(4-Aminophenyl)thiomorpholine-1,1-dioxide

CAS Number: 105297-10-7 MDL Number: MFCD06797046 Molecular Formula: C10H14N2O2S Molecular Weight: 226.29 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 194


Other Product Names / Synonyms: 4-(1,1-Dioxothiomorpholino)aniline, 4-Thiomorpholinoaniline 1',1'-Dioxide

Formula: C₁₀H₁₄N₂O₂S
MW: 226.29 g/mol
Melting Pt: 194 °C
MDL Number: MFCD06797046
CAS Number: 105297-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee474d1ad4c6f5bef82f4b6ff217984039f89017&vid=ee474d1ad4c6f5bef82f4b6ff217984039f89017&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR