MES-Na ≥97.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Sodium 2-MorpholinoethanesulfonateSodium 2-morpholinoethane-1-sulfonateMES-NaSodium 3-morpholin-1-ylethylsulfonate2-(N-Morpholino)ethanesulphonic acid sodium saltSodium 3-morpholin-1-ylethylsulphonate2-(N-Morpholino)ethanesulfonic acid sodium saltSodium 2-morpholinoethane-1-sulphonateSodium 2-Morpholinoethanesulphonate

M0737-25G
TCM0737-025GEA 98.42 USD
TCM0737-025G
MES-Na ≥97.0% (by total nitrogen basis)
MES sodium salt

CAS Number: 71119-23-8
MDL Number: MFCD00065473
Molecular Formula: C6H13NO4S
Molecular Weight: 217.21
Purity/Analysis Method: 97.0% (N)
Form: Crystal


Other Product Names / Synonyms: MES Sodium Salt, 2-Morpholinoethanesulfonic Acid Sodium Salt

Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2
https://us.vwr.com/stibo/web/std.lang.all/42/13/28974213.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR