Myrcene ≥75.0% stabilized
Supplier: TCI America
Synonyms:
7-Methyl-3-methyleneocta-1,6-diene
(stabilized with BHT)
CAS Number: 123-35-3
MDL Number: MFCD00008908
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 37
Specific Gravity (20/20): 0.80
Storage Temperature: 0-10°C
Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: 7-Methyl-3-methylene-1,6-octadiene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 167 °C Density: 0.80 (20 °C) Flash Pt: 37 °C |
MDL Number:
MFCD00008908 CAS Number: 123-35-3 EINECS: 204-622-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...