Methyl lignocerate ≥98.0%

Supplier: TCI America

Synonyms: Lignoceric Acid Methyl EsterMethyl TetracosanoateTetracosanoic Acid Methyl Ester

L0112-1G L0112-5G
TCL0112-005GEA 453.55 USD
TCL0112-005G TCL0112-1G
Methyl lignocerate ≥98.0%
Methyl lignocerate

CAS Number: 2442-49-1
MDL Number: MFCD00009351
Molecular Formula: C25H50O2
Molecular Weight: 382.67
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 60


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Lignoceric Acid Methyl Ester, Methyl Tetracosanoate, Tetracosanoic Acid Methyl Ester

Formula: C₂₅H₅₀O₂
MW: 382.67 g/mol
Boiling Pt: 232 °C
Melting Pt: 60 °C
MDL Number: MFCD00009351
CAS Number: 2442-49-1
https://us.vwr.com/stibo/web/std.lang.all/30/91/28973091.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR