α-Linolenic acid ≥70.0%

Supplier: TCI America

Synonyms: 9Z,12Z,15Z-Octadecatrienoic acid cis,cis,cis-9,12,15-Octadecatrienoic acid Linolenic acid ALA

L0050-25ML
TCL0050-025MLEA 153.2 USD
TCL0050-025ML
α-Linolenic acid ≥70.0%
α-Linolenic acid

CAS Number: 463-40-1
MDL Number: MFCD00065720
Molecular Formula: C18H30O2
Molecular Weight: 278.44
Purity/Analysis Method: 70.0% (GC)
Form: Clear Liquid
Boiling point (°C): 232
Melting point (°C): -11
Flash Point (°C): 110
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids, Regenerative Medicine Research Reagents, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: 9,12,15-Octadecatrienoic Acid

Formula: C₁₈H₃₀O₂
MW: 278.44 g/mol
Boiling Pt: 232 °C
Melting Pt: –11 °C
Density: 0.91 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00065720
CAS Number: 463-40-1
EINECS: 207-334-8
Merck Index: 12,05530
https://us.vwr.com/stibo/web/std.lang.all/30/53/28973053.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR