Kanamycin sulfate ≥94.0% (by total nitrogen basis)

Supplier: TCI America

K0047-25G K0047-5G
TCK0047-25GEA 232.89 USD
TCK0047-25G TCK0047-5G
Kanamycin sulfate ≥94.0% (by total nitrogen basis)
Kanamycin sulphate

CAS Number: 25389-94-0 MDL Number: MFCD00070253 Molecular Formula: C18H36N4O11 Molecular Weight: 582.58 Purity/Analysis Method: 94.0% (N) Form: Crystal Melting point (°C): 250 Specific rotation [a]20/D: 117.5 deg (C=1, H2O)


Product Categories: Bioscience, Glycosides, Sugars (Others), Antibiotic Ingredients Other Product Names / Synonyms: O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1-6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1-4)]-2-deoxy-D-streptamine Monosulfate

Formula: C₁₈H₃₈N₄O₁₅S
Melting Pt: 250 °C
MDL Number: MFCD00070253
CAS Number: 25389-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bfb359d7a08fa707472bab1e0a134cf2a5dbfc8&vid=5bfb359d7a08fa707472bab1e0a134cf2a5dbfc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR