N-Isopropylcyclohexylamine ≥98.0%

Supplier: TCI America

I0373-500ML I0373-25ML
TCI0373-500MLEA 699.5 USD
TCI0373-500ML TCI0373-025ML
N-Isopropylcyclohexylamine ≥98.0%
N-Isopropylcyclohexylamine

CAS Number: 1195-42-2 MDL Number: MFCD00003831 Molecular Formula: C9H19N Molecular Weight: 141.26 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 174 Flash Point (°C): 46 Specific Gravity (20/20): 0.84

Formula: C₉H₁₉N
Boiling Pt: 174 °C
Density: 0.84 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00003831
CAS Number: 1195-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a6f93a7e1f5c114be2f4381c8701e5aefaca4e7&vid=7a6f93a7e1f5c114be2f4381c8701e5aefaca4e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR