Ethyl glycidyl ether ≥98.0%
Supplier: TCI America
CAS Number: 4016-11-9
MDL Number: MFCD00085264
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1-Ethoxy-2,3-epoxypropane, 1,2-Epoxy-3-ethoxypropane
Formula:
C₅H₁₀O₂ Boiling Pt: 128 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00085264 CAS Number: 4016-11-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...